Learn More
Air Systems™ Cascade System: Breathing Air Assembly
Cascade system offers an alternative to air cylinder refill using air compressors
$940.00 - $1009.00
Specifications
Type Breathing Air Assembly
For Use With (Equipment) Self-contained breathing apparatus (SCBA)
Includes Regulator, 4 outlet manifold assembly, low pressure alarm, whistle, relief valve, connecting whip with check valves
Quantity 1 Ea.
View More Specs
Includes: Breathing air assembly: regulator, four-outlet manifold assembly, low pressure whistle, relief valve, and either CGA-346 or CGA-347 connection whips with check valves.
Products 2
Catalog Number Mfr. No. Pressure Price Quantity & Availability  
Catalog Number Mfr. No. Pressure Price Quantity & Availability  
19-806-338
View Documents
Air Systems™
CBA2347NB
4500 psi
Each for $1,009.00
Add to cart
19-120-1209
View Documents
Air Systems™
CBA3346NB
3000 psi
Each for $940.00
Add to cart
Description
Description
  • Air“cascades” across a bank of large supply cylinders to the smaller cylinder being refilled
  • Three-cylinder bottle fill assembly comes with either low or high, 2400 or 5000psi rated fill whip and connecting whips
  • Separate assembly is also available for the purposes of connecting cylinders in series for direct use as breathing air
Order Info
Supply cylinders are not included for either system type and must be purchased separately to complete the apparatus. Optional fill pressure adapter with safety relief valve set at 2400psi and CGA-346, CGA-347, and CGA-702 connection available.
Specifications
Specifications
Breathing Air Assembly
Self-contained breathing apparatus (SCBA)
Regulator, 4 outlet manifold assembly, low pressure alarm, whistle, relief valve, connecting whip with check valves
1 Ea.