Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Air Systems™ Cascade System: Fill Assembly
Cascade system offers an alternative to air cylinder refill using air compressors
Manufacturer:  Air Systems™ CBA3347NB
 View more versions of this product
Catalog No. 19-120-1210
Includes: Breathing air assembly: regulator, four-outlet manifold assembly, low pressure whistle, relief valve, and either CGA-346 or CGA-347 connection whips with check valves.
Description
Description
  • Air“cascades” across a bank of large supply cylinders to the smaller cylinder being refilled
  • Three-cylinder bottle fill assembly comes with either low or high, 2400 or 5000psi rated fill whip and connecting whips
  • Separate assembly is also available for the purposes of connecting cylinders in series for direct use as breathing air
Order Info
Supply cylinders are not included for either system type and must be purchased separately to complete the apparatus. Optional fill pressure adapter with safety relief valve set at 2400psi and CGA-346, CGA-347, and CGA-702 connection available.
Specifications
Specifications
Each
3 cylinder cascade system
Regulator, 4 outlet manifold assembly, low pressure alarm, whistle, relief valve, connecting whip with check valves
Self contained breathing apparatus
Fill Assembly
5000psi