Air Systems CGA Connecting Accessories: Regulators
Manufacturer: Air Systems InternationalSupplier Diversity Partner RG3000
 View more versions of this product
Catalog No. 19-806-405
  Check Availability
Add to Cart
    Specifications
    For Use With (Equipment) Air system cga connecting
    Product Type Adjustable Regulator
    Includes Dual gauge, relief valve, 0.25 in. o.d fitting
    Pressure 3000psig
  • Description & Specifications
Related Products
Product Content Feedback