Learn More
Air Systems™ COOL-BOX™ Breathing Air Compressor Personal Cooling Device
Cools supplied air for supplied-air respirators
Manufacturer:  Air Systems™ BACB196LP
 View more versions of this product
Catalog No. 19-806-308
Add to cart
Description
Description
Easy to operate, low-cost method for cooling supplied air for supplied-air respirators, greatly increasing comfort of workers. Watertight polycarbonate case contains an inner coil covered with ice through which the supplied air passes. Ice cools air from 82°C to 10°C (180°F to 50°F).
  • Places air distribution components closer to work area
  • Unit meets NIOSH point of attachment for supplied-air respirators
  • Cool-Box case has a recessed pull handle and wheels for easy mobility
Model BACB196LP
  • Low-pressure unit for use with Air Systems Ambient Air Pumps
  • Maximum inlet pressure: 20psi
  • 1/2 in. (13mm) Chicago-style inlet fitting
  • 4 respirator outlets with 0.25 in. (6.35mm) Industrial Interchange (Hansen-compatible) quick-disconnect couplers
  • Includes a BACB196K connection kit
Model BACB196
  • High-pressure unit
  • Maximum inlet pressure: 125psi
  • 0.25 in. (6.35mm) male plug inlet fitting (Hansen)
  • 4 respirator outlets witH Industrial Interchange (Hansen-compatible) quick-disconnect couplers
Hoses are not suitable for connecting directly to respirators
Order Info
Hoses sold separately.
Specifications
Specifications
NIOSH
Low Pressure Air Distribution Manifold
Cooling supplied air for supplied air respirator, greatly increasing comfort of workers
20psi
4 hr.
Safety
Air system ambient air pump
4 respirator outlets with 0.37 in., industrial interchange (hansen-compatible) quick-disconnect couplers, 0.5 in. chicago-style inlet fitting, bacb196k connection kit
75cfm
82° to 10°C