Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Air Systems™ COOL-BOX™ Breathing Air Compressor Personal Cooling Device

$761.00 - $788.00

Chemical Identifiers

CAS 7664-38-2
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.994
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
Synonym acide phosphorique
PubChem CID 1004
ChEBI CHEBI:26078
IUPAC Name phosphoric acid
SMILES OP(=O)(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
19-806-307 View Documents Air Systems™
BACB196
Each for $761.00
Add to cart
 
19-806-308 View Documents Air Systems™
BACB196LP
Each for $788.00
Add to cart
 
Description

Description

Easy to operate, low-cost method for cooling supplied air for supplied-air respirators, greatly increasing comfort of workers. Watertight polycarbonate case contains an inner coil covered with ice through which the supplied air passes. Ice cools air from 82°C to 10°C (180°F to 50°F).

  • Places air distribution components closer to work area
  • Unit meets NIOSH point of attachment for supplied-air respirators
  • Cool-Box case has a recessed pull handle and wheels for easy mobility

Model BACB196LP

  • Low-pressure unit for use with Air Systems Ambient Air Pumps
  • Maximum inlet pressure: 20psi
  • 1/2 in. (13mm) Chicago-style inlet fitting
  • 4 respirator outlets with 0.25 in. (6.35mm) Industrial Interchange (Hansen-compatible) quick-disconnect couplers
  • Includes a BACB196K connection kit
Model BACB196
  • High-pressure unit
  • Maximum inlet pressure: 125psi
  • 0.25 in. (6.35mm) male plug inlet fitting (Hansen)
  • 4 respirator outlets witH Industrial Interchange (Hansen-compatible) quick-disconnect couplers

Hoses are not suitable for connecting directly to respirators

Order Info

Hoses sold separately.

Specifications

Chemical Identifiers

7664-38-2
97.994
acide phosphorique
CHEBI:26078
OP(=O)(O)O
H3O4P
NBIIXXVUZAFLBC-UHFFFAOYSA-N
1004
phosphoric acid

Specifications

NIOSH
Cooling supplied air for supplied air respirator, greatly increasing comfort of workers
75cfm
82° to 10°C
Airline type-c mask or hood style respirator
4 respirator outlets with 0.37 in., industrial interchange (hansen-compatible) quick-disconnect couplers, 0.5 in. male plug inlet fitting (hansen)
4 hr.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit