Learn More
Air Systems™ Drag-Bag™
Ideal for rapid intervention and rescue scenarios.
Manufacturer:  Air Systems™ DRAGBAG
 View more versions of this product
Catalog No. 19-500-116
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Includes: Drag-Bag and Drag-Reg regulator/cylinder frame assembly; rescue system includes bag, regulator assembly, and quick-fill hose.
Description
Description
Multi-compartment, denier nylon bag can be used to drag or carry essential respirator components and rescue supplies into difficult rescue/rapid intervention situations.
  • D-ring slots located at each end of the bag allow rope to pull out during search operations and then tie off to the victim during retrieval
  • Side hose compartment includes tube through which hose can be fed into large central (cylinder) compartment
  • Foam padding adds shock resistance while reflective side stripes enhance visibility
  • Shoulder strap and glove handles with wring slots on each bag end for portability
  • All compartments feature dual-zipper closure
Order Info
Bag without regulator/cylinder frame assembly measures 36L x 7W x 10 in.H (91.4 x 17.8 x 25.4cm). Optional accessories available including: 3/8 in. breathing air hose in 15 and 25 ft. (4.6 or 7.6m) coils with “Y“ fittings without couplings and Quick-fill hose, 5 ft. long (1.5m), with female FD-17 coupler with 1/4 male thread, 4500psi.