Learn More
Air Systems™ Low-Pressure Portable Breathing Air Compressors
Portable, oilless sources for use with Type C continuous-flow respirators only (0 to 15psi)
Manufacturer:  Air Systems InternationalSupplier Diversity Partner BAC20P
 View more versions of this product
Catalog No. 19-034-654
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Type c continuous-flow respirator
Ideal to operate 2 hood style respirator or 4 tight fitting constant flow mask
0 to 30psi
46.72kg
CSA
115/208 to 230V 60Hz
60Hz
Ambient Air Pump
0.5 to 0.7μm inlet and compressor filter, on/off switch and plug
115/208 to 230V
103 lb.
20/10A
2hp