Learn More
Air Systems™ MACK-NFPA™ Series Air-Distribution System
Designed for confined-space rescue teams
Manufacturer:  Air Systems™ MACKNFPA3
Catalog No. 19-120-341
Add to cart
Includes: A regulator isolation valve to allow one or both air sources to be used simultaneously, and a CGA-347 high-pressure inlet.
Description
Description
Multi-Air Command Kit (MACK™) for fire/rescue teams working under NFPA guidelines.
  • Dual independent regulators with three-outlet manifold, dual low-pressure alarms, and four 5 ft. long (1.5m) SCBA connect whips
  • In the event of regulator malfunction, rescue can be performed on the standby regulator system
Specifications
Specifications
Fire or rescue teams working under nfpa guidelines
A regulator isolation valve to allow one or both air sources to be used simultaneously, and a cga-347 high-pressure inlet