Learn More
Air Systems™ Replacement Bags for HEPA Vacuums
SDP
For use with AV-2 Series Electric HEPA filtered vacuums
Manufacturer:  Air Systems™ AV2B
Catalog No. 19-821-850
Add to cart
Description
Description
Specifications
Specifications
Paper recovery bag
7600 mL
HEPA
HEPA filter system
256 oz.
Yes
Designed for use on toxic dusts, including Lead, asbestos, nuclear particulate