Learn More
Air Systems™ Venturi-Style Pneumatic Air Blowers
Manufacturer:  Air Systems™ ASI2900
 View more versions of this product
Catalog No. 18-503
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Aluminum base and galvanized steel horn. Expel explosive and hazardous vapors or fumes from underground spaces. No moving parts—horns operate on compressed air or steam. Horn 18-503 meets ventilation requirements for underground storage tanks. Static grounding lug is standard on all models. Horns stand on lightweight aluminum base.
Specifications
Specifications
Venturi pneumatic air blower
44 in.
13 in.
76cfm at 50PSIG
112.4cm