Learn More
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
Syringe Caps
For capping off and sealing any Luer lock or Luer slip type syringe
Manufacturer:  AIR TITE PRODUCTS CO INC STC
 View more versions of this product
Catalog No. 14-817-50
Add to cart
Specifications Description & Specifications
Specifications
Fits all luer-slip and luer-lock syringes, laboratory and veterinary use
Polypropylene
Sterile
Syringe Cap