Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Air Track Accessories
Use with air track (S52229) to study linear motion.
$155.00
Catalog Number | Mfr. No. | Product Type | Length (English) | Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Length (English) | Length (Metric) | Price | Quantity & Availability | |||
S85363
![]() |
Science First![]() 12515 |
Glider set | N/A | N/A |
Each for $155.00
|
|
Description
Bumpers, magnets, and dashpot simply plug into the gliders as needed.
Lab Manual
- Contains instructions, suggestions for experiments, and activities
- Four 105g masses
- Screws
- Wingnuts
- One anodized glider: 3 in. (7.5cm), 3.7 oz. (105g)
- One anodized glider: 5.9 in. (15cm), 7.4 oz. (210g)
- Four small bumpers
- Four large bumpers
- One set mounted magnets for elastic collisions
- One set dashpot and coupler for inelastic collisions
- Two weak springs
- One pulley
- One hook and loop tape set
Specifications
Air track (S52229) |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title