Learn More
AirClean™ Systems PowderSafe™ Type C Balance Enclosures
Thermally-fused polypropylene makes the Airclean PowderSafe™ Type C Balance Enclosure suitable for weighing powders and/or solvents.
$9809.00 - $15998.00
Specifications
For Use With (Application) For weighing powders/solvents
Frame Material Polypropylene
Lighting Vapor-proof fluorescent light
Product Type Type C Balance Enclosure
View More Specs
Products 6
Catalog Number Mfr. No. Dimensions (D x W x H) Interior Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (D x W x H) Interior Price Quantity & Availability  
36-101-8906
View Documents
AirClean™ Systems
AC760C
58.4 x 89 x 63.5 cm
Each for $11,402.00
Add to cart
36-101-8908
View Documents
AirClean™ Systems
AC770C
58.4 x 119.3 x 63.5 cm
Each for $11,754.00
Add to cart
36-101-8907
View Documents
AirClean™ Systems
AC765C
58.4 x 89 x 89 cm
Each for $9,809.00
Add to cart
36-101-8909
View Documents
AirClean™ Systems
AC775C
55.8 x 119.3 x 89 cm (22 x 47 x 35 in.)
Each for $11,372.00
Add to cart
36-101-8911
View Documents
AirClean™ Systems
AC785C
76.2 x 183 x 137 cm
Each for $15,998.00
Add to cart
36-101-8910
View Documents
AirClean™ Systems
AC780C
58.4 x 180.3 x 63.5 cm
Each for $13,766.00
Add to cart
Description
Description
  • AirSafe™ automatic safety controller
  • Horizontal laminar airflow pattern
  • Chemically impervious construction
  • Available in 36 in., 48 in., and 72™ widths
  • HEPASafe filter change out
  • High mass - solid polypropylene construction
  • Vapor-proof fluorescent light
  • ASHRAE 110 Tested
Options:
  • Dark blue base
  • Bonded carbon multipurpose filtration
  • Waste port for weighing vessels
Specifications
Specifications
For weighing powders/solvents
Polypropylene
Vapor-proof fluorescent light
Type C Balance Enclosure