Learn More
Airclean™ System AC600 Series Ductless Endoscopy Workstations
Provide mobile solution for operator protection from high-level disinfectant vapors during endoscope reprocessing
Manufacturer:  AIRCLEAN SYSTEMS AC648C
 View more versions of this product
Catalog No. 36-100-0008
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
500 cfm
Glutaraldehyde, OPA, Cidex™, Peracetic Acid, Hydrogen Peroxide
22 in.
55.88cm
32 in.
ANSI Z9.5, A.A.M.I. ST58, OSHA and JCAHO
A mobile solution for operator protection from disinfectant fumes
81.28cm
110/220V
48 in.