Learn More
AirClean™ Systems Polypropylene Hood Blowers
SDP
Used especially in situations that require resistance to chemical agents, silent operation and energy savings
Supplier:  AirClean™ Systems VSB30
 View more versions of this product
Catalog No. 36-100-0038
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Alert:
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Description
Description
  • Injection-molded
  • Resistant to UV rays and chemical agents
  • Adjusted to eight positions through 360°
  • Constructed for outdoor use
Specifications
Specifications
8-position Setting through 360°
11.81 in.
Situations that require resistance to chemical agents, silent operation, energy savings
11.81 in.
30 cm
Product Suggestions
Product Suggestions