Airclean™ System TEE Probe Storage Cabinets
Designed to accommodate up to three disinfected probes
Manufacturer: AIRCLEAN SYSTEMS ACTE06
 View more versions of this product
Catalog No. 36-100-0013
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Material Thermally fused polypropylene
    For Use With (Application) Designed for TEE probes
    Height (English) Exterior 88 in.
    Height (Metric) Exterior 223.52cm
    Width (English) Exterior 51 in.
  • Description & Specifications
Related Products
Product Content Feedback