AirClean™ Systems AirClean™ Systems Activated Bonded Carbon Chemisorptive Filters
For the removal of fumes and vapors
Manufacturer: AirClean™ Systems ACF400
 View more versions of this product
Catalog No. 36-101-8828
  Check Availability
Add to Cart
    Specifications
    Filter Type ACF400
    Material Activated Carbon
    For Use With (Application) Inorganic acids
  • Description & Specifications
Related Products
Product Content Feedback