Learn More
AirClean™ Systems Activated Bonded Carbon Chemisorptive Filters
SDP
For the removal of fumes and vapors
Supplier:  AirClean™ Systems ACFFOR
 View more versions of this product
Catalog No. 36-101-8829
Add to Cart
Description
Description
  • Coconut shell activated carbon
  • Chemically impregnated carbon for increased load capacity
  • Large surface area and highly developed internal structure allows adsorption up to 50%
  • Bonded design eliminates carbon dusting and prevents carbon shift that can lead to “dead spots”
Specifications
Specifications
ACFFOR
Activated Carbon
Formaldehyde
Product Suggestions
Product Suggestions