Learn More
AirClean Systems Activated Bonded Carbon Chemisorptive Filters
For the removal of fumes and vapors
Manufacturer: AirClean Systems ACFGLT
Description
- Coconut shell activated carbon
- Chemically impregnated carbon for increased load capacity
- Large surface area and highly developed internal structure allows adsorption up to 50%
- Bonded design eliminates carbon dusting and prevents carbon shift that can lead to “dead spots”
Specifications
ACFGLT | |
Glutaraldehyde | |
Activated Carbon |