Learn More
AirClean™ Systems Bulk Powder Handling Enclosures
SDP
Provide effective control and containment of airborne particulate during manipulation and transfer of potent compounds
Supplier:  AirClean™ Systems AC795BE
 View more versions of this product
Catalog No. 36-101-8901
Add to Cart
Description
Description
  • Designed to provide turbulent free airflow environment
  • Air moves in horizontal pattern to maximize containment while minimizing sample loss and balance instability
  • HEPASafe™ filter change-out technology allows operator to safely and effectively change both pre-filter and primary HEPA filter
  • Preset face velocity that is constantly monitored and maintained by AirSafe™ automatic safety controller
  • Audible and visible alarms when filter change out is necessary
  • Optional bonded carbon filtration for removal of fumes during bulk chemical handling and dispensing is also available
  • With sturdy stand which provides comfortable working height of 39 in.
  • Available in 4 ft., 5 ft. and 6 ft. widths with nominal depth of 36 in.
Specifications
Specifications
Bulk Powder Handling Enclosure
Product Suggestions
Product Suggestions