Combination PCR Workstation
The Airclean Combination PCR Workstation combines ISO 5 (FS209E) Class 100 clean air environment with UV light sterilization for optimal protection from sample contamination.
Manufacturer: AIRCLEAN SYSTEMS AC632TLFUVC
 View more versions of this product
Catalog No. 36-101-8895
 N/A
N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Product Type Combination PCR Workstation
    Dimension (English) Interior 23D x 31W x 27H in.
    Dimension (Metric) Interior 58.4D x 78.7W x 68.5H cm
    Lighting Digital UV light
  • Description & Specifications
Related Products
Product Content Feedback