Learn More
Combination PCR Workstation
The Airclean Combination PCR Workstation combines ISO 5 (FS209E) Class 100 clean air environment with UV light sterilization for optimal protection from sample contamination.
Manufacturer:  AIRCLEAN SYSTEMS AC648TLFUVC
 View more versions of this product
Catalog No. 36-101-8896
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
23D x 47W x 29H in.
58.4D x 119.3W x 73.6H cm
Combination PCR Workstation
Digital UV light