Learn More
AirClean™ Systems Dead Air Boxes
Provides circulation free environment with UV light sterilization
Manufacturer:  AIRCLEAN SYSTEMS AC624DB
 View more versions of this product
Catalog No. 36-101-8888
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Dead Air Box
58.4D x 58.4W x 53.3H cm
Safety switch to prevent exposure to harmful UV light; Full access folding sash; UV and fluorescent lights; PP ans PC unibody
23D x 23W x 21H in.
Ductless enclosures