AirClean™ Systems Dead Air Boxes
Provides circulation free environment with UV light sterilization
Manufacturer: AIRCLEAN SYSTEMS AC632DB
 View more versions of this product
Catalog No. 36-101-8889
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Dimension (English) Interior 23D x 31W x 21H in.
    Dimension (Metric) Interior 58.4D x 78.7W x 53.3H cm
    For Use With (Equipment) Ductless enclosures
    Includes Safety switch to prevent exposure to harmful UV light; Full access folding sash; UV and fluorescent lights; PP ans PC unibody
    Type Dead Air Box
  • Description & Specifications
Related Products
Product Content Feedback