AirClean™ Systems Dead Air Boxes
Provides circulation free environment with UV light sterilization
Manufacturer: AIRCLEAN SYSTEMS AC648DB
 View more versions of this product
Catalog No. 36-101-8891
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Dimension (English) Interior 23D x 47W x 23H in.
    Dimension (Metric) Interior 58.4D x 119.3W x 58.4H cm
    Includes Safety switch to prevent exposure to harmful UV light; Full access folding sash; UV and fluorescent lights; PP ans PC unibody
    For Use With (Equipment) Ductless enclosures
    Type Dead Air Box
  • Description & Specifications
Related Products
Product Content Feedback