Learn More
All-Plastic Norm-Ject™ Syringes
Inert syringes for use in gas or liquid chromatography, in vitro fertilization (IVF), embryo-transfer, and various laboratory procedures. Air-Tite All Plastic Norm-Ject syringes are available compatible with a variety of needle styles. Needles are sold separately.
Manufacturer:  AIR TITE PRODUCTS CO INC A50
 View more versions of this product
Catalog No. 14-817-35
Due to product restrictions, please to purchase or view availability for this product.
Requires : Purchase needles separately.
Specifications
Description
  • Needle tips not included and must be purchased separately
  • Graduated syringes manufactured from high-quality laboratory-grade polypropylene and polyethylene
  • Chemically inert and noncytotoxic, these syringes are ideal for chromatography, in vitro fertilization and embryo transfer
  • Luer Tip, Luer-Slip, or catheter tip compatibility
  • Polypropylene barrel and polyethylene plunger are chemically inert and noncytotoxic; needles contain no latex, synthetic rubber, silicone oil, or styrene
  • Positive safety stop prevents accidental spills
  • FDA registered
Description & Specifications
Specifications
Yes
Cone
FDA, CE, ISO 9001, DEHP Free
Luer Tip
Yes
Eccentric, luer-tip
Norm-Ject™ Syringe
50mL
For nuclear medicine, amniocentisis, IVF, embryo-transfer, chromatography and many laboratory procedures
1cc
Plastic
50.0mL
Accessories