Learn More
Brady™ Aisle Marking Tape Arrows - Adhesive Vinyl, Solid Color, Orange
Permanent acrylic adhesive sticks to a wide variety of surfaces
Manufacturer:  Brady™ 121451
 View more versions of this product
Catalog No. 19-103-720
Add to cart
Description
Description
  • Indoor/outdoor vinyl film withstands exposure to moisture, chemicals and handling
  • Ideal for marking aisles, passageways and storage locations and identifying equipment and egress paths
Specifications
Specifications
No Text
Safety
Vinyl
2 in.
2 in.
Pack of 100 Each
Orange
Marking Tape
0.004 in.