Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AK2 Mouse anti-Human, Alexa Fluor™ 750, Clone: AT7E7, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250575AF750
Description
AK2 Monoclonal specifically detects AK2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
AK2 | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
AK2 | |
Recombinant human AK2 (1-239aa) purified from E. coli | |
100 μL | |
204.0 | |
Store at 4°C in the dark. |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
AT7E7 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
adenylate kinase 2, adenylate kinase 2, mitochondrial, adenylate kinase isoenzyme 2, mitochondrial, ADK2, AK 2, ATP-AMP transphosphorylase 2, EC 2.7.4, EC 2.7.4.3 | |
Mouse | |
Protein A purified | |
Primary | |
Human | |
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title