Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AK3L1 Mouse anti-Human, Mouse, Rat, Canine, Monkey, Allophycocyanin/Cy7, Clone: OTI3B1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270129APCCY7

Catalog No. N270129APC7


Add to cart

Description

Description

AK3L1 Monoclonal antibody specifically detects AK3L1 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Flow Cytometry.
Specifications

Specifications

AK3L1
OTI3B1
Flow Cytometry
Affinity Purified, Conjugated
AK4
Full length recombinant protein of human AK3L1 (NP_037542) produced in HEK293 cell.
Immunogen affinity purified
Core ESC Like Genes, Stem Cell Markers
Primary
205.0
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
adenylate kinase 3, Adenylate kinase 3-like, adenylate kinase 3-like 1, adenylate kinase 4, AK3adenylate kinase isoenzyme 4, mitochondrial, AK3L1, AK3L2, ATP-AMP transphosphorylase, EC 2.7.4, EC 2.7.4.3, GTP:AMP phosphotransferase, MGC166959, mitochondrial adenylate kinase-3, nucleoside-triphosphate-adenylate kinase
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Canine, Human, Monkey, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit