Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AK3L1 Mouse anti-Human, Mouse, Rat, Canine, Monkey, HRP, Clone: OTI3B1, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP270129H
Description
AK3L1 Monoclonal antibody specifically detects AK3L1 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.Specifications
AK3L1 | |
OTI3B1 | |
Western Blot, Immunohistochemistry | |
Affinity Purified, Conjugated | |
AK4 | |
Full length recombinant protein of human AK3L1 (NP_037542) produced in HEK293 cell. | |
Immunogen affinity purified | |
Core ESC Like Genes, Stem Cell Markers | |
Primary | |
205.0 |
Immunohistochemistry, Western Blot | |
HRP | |
PBS with No Preservative | |
adenylate kinase 3, Adenylate kinase 3-like, adenylate kinase 3-like 1, adenylate kinase 4, AK3adenylate kinase isoenzyme 4, mitochondrial, AK3L1, AK3L2, ATP-AMP transphosphorylase, EC 2.7.4, EC 2.7.4.3, GTP:AMP phosphotransferase, MGC166959, mitochondrial adenylate kinase-3, nucleoside-triphosphate-adenylate kinase | |
Mouse | |
IgG1 | |
0.1 mL | |
Store at 4°C in the dark. | |
Monoclonal | |
Canine, Human, Monkey, Mouse, Rat |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title