Learn More
Abnova™ AK3L2 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00000205T02
Catalog No. 89-932-955
Add to cart
Description
Description
Specifications
Specifications
293T
NM_001002921
pCMV-AK3L2 full-length
100 μL
Human
AK3L2 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 25.3kDa
Transient overexpression cell lysate was tested with anti-AK3L2 antibody (Mfr. No. H00000205-B02) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot