Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AK5 Mouse anti-Human, PE/Atto594, Clone: OTI2F9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271813PEATT594

Catalog No. N271813P594


Add to cart

Description

Description

AK5 Monoclonal antibody specifically detects AK5 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

AK5
OTI2F9
Flow Cytometry
Affinity Purified, Conjugated
AK5
Full length recombinant protein of human AK5 (NP_777283) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Flow Cytometry
PE/ATTO 594
PBS with 0.05% Sodium Azide
adenylate kinase 5, adenylate kinase 6, adenylate kinase isoenzyme 5, AK 5, AK6, ATP-AMP transphosphorylase 5, EC 2.7.4.3, MGC33326
Mouse
IgG2b
0.1 mL
Primary
26289.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit