Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AKAP8L Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 101031AP150UL
Description
Could play a role in constitutive transport element (CTE)-mediated gene expression. Does not seem to be implicated in the binding of regulatory subunit II of PKA. May be involved in nuclear envelope breakdown and chromatin condensation. May regulate the initiation phase of DNA replication when associated with TMPO-beta.Specifications
AKAP8L | |
Polyclonal | |
Unconjugated | |
AKAP8L | |
A kinase anchor protein 8 like, AKAP8 like protein, AKAP8L, HA95, HAP95, Helicase A binding protein 95, Homologous to AKAP95 protein, NAKAP, NAKAP95, Neighbor of AKAP95 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26993 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9ULX6 | |
AKAP8L | |
AKAP8L Fusion Protein Ag0145 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction