Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AKAP9 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518345
Description
This antibody is predicted to react with mouse based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
The A-kinase anchor proteins are a group of structurally diverse proteins which have the common function of binding to the regulatory subunit of protein kinase A and confining the holoenzyme to discrete locations within the cell. This gene encodes a member of the AKAP family. Alternate splicing of this gene results in at least two isoforms that localize to the centrosome and the Golgi apparatus, and interact with numerous signaling proteins from multiple signal transduction pathways. These signaling proteins include type II protein kinase A, serine/threonine kinase protein kinase N, protein phosphatase 1, protein phosphatase 2a, protein kinase C-epsilon and phosphodiesterase 4D3.
Specifications
AKAP9 | |
Polyclonal | |
Unconjugated | |
AKAP9 | |
5730481H23Rik; A kinase (PRKA) anchor protein (yotiao) 9; A kinase (PRKA) anchor protein 9; AKAP 120-like protein; AKAP 350; AKAP 450; AKAP350; AKAP450; AKAP9; AKAP-9; AKAP9-BRAF fusion protein; A-kinase anchor protein 350 kDa; A-kinase anchor protein 450 kDa; A-kinase anchor protein 9; A-kinase anchoring protein 9; AW545847; C79026; centrosome- and Golgi-localized PKN-associated protein; centrosome- and golgi-localized protein kinase N-associated protein; CG-NAP; G1-448-15; HYPERION; KIAA0803; kinase N-associated protein; LQT11; mei2-5; mKIAA0803; MU-RMS-40.16A; PPP1R45; PRKA9; Protein hyperion; protein kinase A anchoring protein 9; protein kinase A-anchoring protein 9; protein phosphatase 1, regulatory subunit 45; protein yotiao; repro12; YOTIAO | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
10142 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q99996 | |
AKAP9 | |
Synthetic peptide sequence (SGSTTQFHAGMR) corresponding to the C-terminus amino acids of AKAP9 (aa 3895-3907). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction