Akers Biosciences™ BreathScan™ Alcohol Detector
Fast, easy and portable way to help detect the presence of alcohol in an individual's breath
Manufacturer: AKERS BIOSCIENCES INC 4000128
 View more versions of this product
Catalog No. 22-740-126
  Check Availability
Add to Cart
Alert
Only for use as a screening device and indication of the possible presence of alcohol in the test subject's blood.
    Specifications
    Product Type BreathScan Alcohol detector
    Detectable Analytes 0.08% Alcohol
    Quantity 100 Pack
    Test Time 2 min.
    Certifications/Compliance FDA
  • Description & Specifications
Related Products
Product Content Feedback