Learn More
Akers Biosciences™ BreathScan™ Alcohol Detector
Fast, easy and portable way to help detect the presence of alcohol in an individual's breath
Manufacturer:  AKERS BIOSCIENCES INC 4000122
 View more versions of this product
Catalog No. 22-740-124
  Check Availability
Add to cart
Alert:
Only for use as a screening device and indication of the possible presence of alcohol in the test subject's blood.
Specifications Description & Specifications
Specifications
FDA
100 Pack
BreathScan Alcohol detector
0.02% Alcohol
2 min.