Akers Biosciences™ PIFA™ Heparin/PF4 Rapid Assay
Single-use, single patient test that can easily be performed in approximately 10 minutes
Manufacturer: AKERS BIOSCIENCES INC 4036025
 View more versions of this product
Catalog No. 22-740-120
  Check Availability
Add to Cart
    Specifications
    Product Type Heparin/PF4 Rapid Assay Kit
    For Use With (Application) Antibody related to heparin-induced thrombocytopenia
    Includes Easy-to-interpret results window, 1 package insert and 6 individual sealed pouches, each pouch contains 1 PIFA™ heparin/PF4 minireactor device
    Quantity 6 Tests
    Test Time ≈10 min.
  • Description & Specifications
Related Products
Product Content Feedback