Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AKNA Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA545800
Description
Peptide sequence: AGGAVTGDPL GPPPADTLQC PLCGQVGSPP EADGPGSATS GAEKATTRRK Sequence homology: Human: 100%; Rabbit: 92%; Rat: 85%.
AKNA is a transcription factor that specifically activates the expression of the CD40 receptor and its ligand CD40L/CD154, two cell surface molecules on lymphocytes that are critical for antigen-dependent-B-cell development. It binds to A/T-rich promoters.
Specifications
AKNA | |
Polyclonal | |
Unconjugated | |
AKNA | |
AKNA; AKNA transcript F2; AT-hook transcription factor; AT-hook transcription factor AKNA; AT-hook-containing transcription factor; KIAA1968; Microtubule organization protein AKNA | |
Rabbit | |
Affinity Chromatography | |
RUO | |
80709 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q7Z591 | |
AKNA | |
Synthetic peptide directed towards the C-terminal of human AKNA (aa 1252-1301). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction