Learn More
Abnova™ AKR1A1 293T Cell Transient Overexpression Lysate (Denatured) (T01)
Human overexpression lysate
Manufacturer:  Abnova™ H00010327T01
Catalog No. 89-933-001
Add to cart
Description
Description
Specifications
Specifications
293T
NM_006066.2
pCMV-AKR1A1 full-length
100 μL
Human
AKR1A1 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 35.86kDa
Transient overexpression cell lysate was tested with anti-AKR1A1 antibody (Mfr. No. H00010327-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot