Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AKR1A1 Mouse anti-Human, Clone: OTI3C7, liquid, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene TA500767
Description
This gene encodes a member of the aldo/keto reductase superfamily, which consists of more than 40 known enzymes and proteins. This member, also known as aldehyde reductase, is involved in the reduction of biogenic and xenobiotic aldehydes and is present in virtually every tissue. Multiple alternatively spliced transcript variants of this gene exist, all encoding the same protein.Specifications
AKR1A1 | |
Monoclonal | |
0.68 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P14550 | |
ALDR1, ALR, ARM, DD3, HEL-S-6 | |
Mouse | |
IgG2a | |
100 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
10327 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI3C7 | |
Unconjugated | |
AKR1A1 | |
Liquid | |
AKR1A1 | |
Full length human recombit protein of human AKR1A1 produced in HEK293T cell. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title