Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKR1A1 Mouse anti-Human, Clone: OTI8E12, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA500741

Catalog No. 50-167-1882


Add to cart

Description

Description

This gene encodes a member of the aldo/keto reductase superfamily, which consists of more than 40 known enzymes and proteins. This member, also known as aldehyde reductase, is involved in the reduction of biogenic and xenobiotic aldehydes and is present in virtually every tissue. Multiple alternatively spliced transcript variants of this gene exist, all encoding the same protein.
Specifications

Specifications

AKR1A1
Monoclonal
0.58 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P14550
ALDR1, ALR, ARM, DD3, HEL-S-6
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10327
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI8E12
Unconjugated
AKR1A1
Liquid
AKR1A1
Full length human recombit protein of human AKR1A1 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit