Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKR1A1 Mouse anti-Human, PE/Cy5.5, Clone: OTI6E3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271591PECY55

Catalog No. N271591PEC5


Add to cart

Description

Description

AKR1A1 Monoclonal antibody specifically detects AKR1A1 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

AKR1A1
OTI6E3
Flow Cytometry
Affinity Purified, Conjugated
AKR1A1
Full length recombinant protein of human AKR1A1 (NP_006057) produced in HEK293T cells.
Immunogen affinity purified
Stem Cell Markers
Primary
10327.0
Flow Cytometry
PE-Cyanine5.5
PBS with 0.05% Sodium Azide
alcohol dehydrogenase, alcohol dehydrogenase [NADP+], Aldehyde reductase, Aldo-keto reductase family 1 member A1, aldo-keto reductase family 1, member A1 (aldehyde reductase), ALDR1, ALRMGC1380, ARM, DD3, dihydrodiol dehydrogenase 3, EC 1.1.1, EC 1.1.1.2, MGC12529
Mouse
IgG2a
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit