Learn More
AKR1A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA549731
Catalog No. PA549731
Add to cart
Description
Description
The antibody detects endogenous level of total AKR1A1 protein.
This gene encodes a member of the aldo/keto reductase superfamily, which consists of more than 40 known enzymes and proteins. This member, also known as aldehyde reductase, is involved in the reduction of biogenic and xenobiotic aldehydes and is present in virtually every tissue. Multiple alternatively spliced transcript variants of this gene exist, all encoding the same protein.
Specifications
Specifications
AKR1A1
Polyclonal
Unconjugated
AKR1A1
Liquid
AKR1A1
Synthetic peptide corresponding to a region derived from 312-324 amino acids of human aldo-keto reductase family 1, member A1 (aldehyde reductase)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
ELISA, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
P14550, Q9JII6, P51635
Aldo-keto reductase family 1, member A1 (aldehyde reductase), ALR, ARM, DD3, ALDR1
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10327, 58810, 78959