Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AKR1B1, Mouse anti-Human, Clone: CPTC-AKR1B1-2, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP27979720UG
Description
AKR1B1 Monoclonal antibody specifically detects AKR1B1 in Human Samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array.Specifications
AKR1B1 | |
CPTC-AKR1B1-2 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Purified | |
AKR1B1 | |
Recombinant full-length human AKR1B1 protein | |
Protein A or G purified | |
RUO | |
Store at 4°C. | |
Monoclonal | |
Human |
Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array, Western Blot | |
0.2 mg/mL | |
Western Blot 1-2 ug/ml, Immunohistochemistry 1-2 ug/ml, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
P15121 | |
ADR, Aldehyde reductase, Aldo-keto reductase family 1 member B1, aldo-keto reductase family 1, member B1 (aldose reductase), ALDR1aldose reductase, ALR2, ARaldehyde reductase 1, EC 1.1.1, EC 1.1.1.21, Lii5-2 CTCL tumor antigen, low Km aldose reductase, MGC1804 | |
Mouse | |
IgG1 Kappa | |
20 ug | |
Stem Cell Markers | |
Primary | |
231 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title