Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AKR1B1, Mouse anti-Human, DyLight 650, Clone: CPTC-AKR1B1-2, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP279931C
Description
AKR1B1 Monoclonal antibody specifically detects AKR1B1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).Specifications
AKR1B1 | |
CPTC-AKR1B1-2 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
Conjugated, Purified | |
AKR1B1 | |
Recombinant full-length human AKR1B1 protein | |
Protein A or G purified | |
RUO | |
Store at 4°C in the dark. | |
Monoclonal |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ADR, Aldehyde reductase, Aldo-keto reductase family 1 member B1, aldo-keto reductase family 1, member B1 (aldose reductase), ALDR1aldose reductase, ALR2, ARaldehyde reductase 1, EC 1.1.1, EC 1.1.1.21, Lii5-2 CTCL tumor antigen, low Km aldose reductase, MGC1804 | |
Mouse | |
IgG1 Kappa | |
0.1 mL | |
Stem Cell Markers | |
Primary | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title