Learn More
Abnova™ AKR1B10 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00057016T01
Catalog No. 89-933-010
Add to cart
Description
Description
Specifications
Specifications
293T
NM_020299.3
pCMV-AKR1B10 full-length
100 μL
Human
AKR1B10 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 36kDa
Transient overexpression cell lysate was tested with anti-AKR1B10 antibody (Mfr. No. H00057016-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot