Learn More
AKR1C3 293T Cell Overexpression Lysate (Denatured), Abnova
Cell transient overexpression lysate for AKR1C3
Manufacturer:  Abnova Corporation H00008644T02
 View more versions of this product
Catalog No. 89-000-339
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Western Blot
293T
36.9
pCMV-AKR1C3 full-length
100μL
-80°C
Human
NM_003739.4
6.8
Transient overexpression cell Lysate was tested with Anti-AKR1C3 antibody (H00008644-B02) by western blotting
1X Sample Buffer: 50mM Tris HCl, 2% SDS, 300mM 2-mercaptoethanol, 0.01% Bromophenol Blue, 10% Glycerol