Learn More
Abnova™ AKR1C4 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00001109T01
Catalog No. 89-933-024
Add to cart
Description
Description
Specifications
Specifications
293T
BC020744.1
pCMV-AKR1C4 full-length
100 μL
Human
AKR1C4 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 35.64kDa
Transient overexpression cell lysate was tested with anti-AKR1C4 antibody (Mfr. No. H00001109-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot