Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AKR7A2 Mouse anti-Human, Fish, Zebrafish, Clone: C10-D5, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP267126
Description
AKR7A2 Monoclonal antibody specifically detects AKR7A2 in Human, Fish, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.Specifications
AKR7A2 | |
C10-D5 | |
Unconjugated | |
Purified | |
AFAR1, AFARAFB1-AR1, AFB1 aldehyde reductase 1, AFB1-AR 1, aflatoxin B1 aldehyde reductase member 2, aflatoxin beta1 aldehyde reductase, AKR7, aldo-keto reductase family 7, member A2 (aflatoxin aldehyde reductase), Aldoketoreductase 7, EC 1.1.1.n11, SSA reductase, Succinic semialdehyde reductase | |
AKR7A2 | |
Peptide | |
Monoclonal | |
Protein A purified | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot,Immunohistochemistry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin) | |
2 mg/mL | |
Western Blot 1:1000-1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:200, Immunohistochemistry-Paraffin | |
TBS (pH7.4), 1%BSA, 40%Glycerol with 0.05% Sodium Azide | |
8574 | |
Mouse | |
IgG1 | |
Primary | |
100μL | |
Human,Fish,Zebrafish |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title