Learn More
Akro-Mils™ Akro-Grid Storage Totes: 0.59 cu. ft./40 lb.
With optional lengthwise and crosswise dividers for smaller storage
Manufacturer:  Akro-Mils™ 33168GREY
 View more versions of this product
Catalog No. 08-757-866
Add to cart
Description
Description
These lightweight totes facilitate work-in-progress transfers; enable compartmentalized storage for items used in concert.
  • Offer hundreds of storage configurations; can be subdivided down to a 1 1/8 in. square
  • Injection-molded from polypropylene block copolymer
  • Resist most solvents and chemicals
  • Choice of Red, Blue or Gray for color-coding and easy identification
  • Available in eight sizes, many with the same footprint for secure stacking
  • Stacking rims and external ribbing add strength
  • Compatible with most racks, shelving and automated material handling equipment
  • Dividers, lids and label holder sold separately
Specifications
Specifications
16.5 x 10.87 x 8 in. (42 x 27.6 x 20.3cm)
0.59 cu. ft.
20.3cm
42cm
27.6cm
14.75 in.
9.3 in.
14.9 in.
16.7L
8 in.
16.5 in.
10.9 in.
Gray
37.4cm
23.6cm
37.8cm